4-(Indol-3-yl)butyric acid 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-Indolebutyric acid, Indole-3-butyric acid
A15260-06
A15260-14
A15260-22
CAAAA15260-06EA
73.79
CAD
CAAAA15260-06
CAAAA15260-14
CAAAA15260-22
4-(Indol-3-yl)butyric acid 98%
4-(Indol-3-yl)butyric acid
Purity: 98%
CAS No.: 133-32-4
Molecular Formula: C12H13NO2
Formula Weight: 203.24
Odor: Odorless
Melting Point: 121-125°
Beilstein Reference: 171120
Merck Reference: 14,4965
UN No.: 2811
MDL No.: MFCD00005664
Formula:
C₁₂H₁₃NO₂ MW: 203.24 Melting Pt: 121…125 °C Density: 1.27 Storage Temperature: Ambient |
MDL Number:
MFCD00005664 CAS Number: 133-32-4 EINECS: 205-101-5 UN: 2811 ADR: 6.1 III Merck Index: 14,4965 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...