α-Bromo-m-toluonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-(Bromomethyl)benzonitrile, m-(Bromomethyl)benzonitrile
A15244-06
A15244-14
CAAAAA15244-22EA
1098.81
CAD
CAAAAA15244-22
CAAAA15244-06
CAAAA15244-14
α-Bromo-m-toluonitrile 98%
α-Bromo-m-toluonitrile
Purity: 98%
CAS No.: 28188-41-2
Molecular Formula: C8H6BrN
Formula Weight: 196.05
Melting Point: 92-96°
Beilstein Reference: 742356
UN No.: 2923
MDL No.: MFCD00001809
Formula:
C₈H₆BrN MW: 196.05 g/mol Boiling Pt: 130 Melting Pt: 92…96 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001809 CAS Number: 28188-41-2 EINECS: 248-890-1 UN: 1759 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...