2-Phenylethanol 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15241-0B
A15241-0I
A15241-30
CAAAA15241-0BEA
167.48
CAD
CAAAA15241-0B
CAAAA15241-0I
CAAAA15241-30
2-Phenylethanol 98+%
2-Phenylethanol
Purity: 98+%
CAS No.: 60-12-8
Molecular Formula: C8H10O
Formula Weight: 122.17
Flash Point: 101°(214°F)
Physical Form: Liquid
Density: 1.021
Refractive Index: 1.5330
Appearance: Colorless
Melting Point: -27°
Boiling Point: 219°
Beilstein Reference: 1905732
Merck Reference: 14,7224
UN No.: 2810
MDL No.: MFCD00002886
Formula:
C₆H₅CH₂CH₂OH Boiling Pt: 219 °C Melting Pt: –27 °C Density: 1.021 Flash Pt: 101 °C(214 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002886 CAS Number: 60-12-8 EINECS: 200-456-2 UN: 2810 ADR: 6.1,III Merck Index: 14,7224 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...