4-Acetylbenzoic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Acetylbenzoic Acid, 4'-Acetophenonecarboxylic acid

A15239-03 A15239-06 A15239-14
CAAAA15239-03EA 62.41 CAD
CAAAA15239-03 CAAAA15239-06 CAAAA15239-14
4-Acetylbenzoic acid 98+%
4-Acetylbenzoic acid

Purity: 98+% CAS No.: 586-89-0 Molecular Formula: C9H8O3 Formula Weight: 164.16 Odor: Odorless Melting Point: 208-212° Beilstein Reference: 2207355 MDL No.: MFCD00002561

Formula: C₉H₈O₃
MW: 164.16 g/mol
Boiling Pt: 340
Melting Pt: 208…212 °C
Storage Temperature: Ambient
MDL Number: MFCD00002561
CAS Number: 586-89-0
EINECS: 209-588-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR