α-Bromo-4-nitrotoluene 97+%

Supplier: Thermo Scientific Chemicals
Danger

A15236-14 A15236-22 A15236-36
CAAAA15236-14EA 69.99 CAD
CAAAA15236-14 CAAAA15236-22 CAAAA15236-36
α-Bromo-4-nitrotoluene 97+%
α-Bromo-4-nitrotoluene

Purity: 97+% CAS No.: 100-11-8 Molecular Formula: C7H6BrNO2 Formula Weight: 216.04 Melting Point: 97-101° Beilstein Reference: 742796 UN No.: 3261 MDL No.: MFCD00007373

Formula: C₇H₆BrNO₂
Boiling Pt: 304 °C (1013 hPa)
Melting Pt: 97…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00007373
CAS Number: 100-11-8
EINECS: 202-820-6
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR