1,3-Diphenylguanidine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N,N'-Diphenylguanidine, sym-Diphenylguanidine

A15234-22 A15234-0B A15234-30
CAAAAA15234-22EA 42.03 CAD
CAAAAA15234-22 CAAAA15234-0B CAAAA15234-30
1,3-Diphenylguanidine 97%
1,3-Diphenylguanidine

MDL: MFCD00001758 Beilstein: 1875653 Soluble in alcohol, chloroform, hot benzene, hot toluene, and dilute mineral acids


Purity: 97% CAS No.: 102-06-7 Molecular Formula: C13H13N3 Formula Weight: 211.27 Flash Point: 170°(338°F) Odor: Characteristic Boiling Point: 170° dec. Solubility: Soluble in alcohol, chloroform, hot benzene, hot toluene, and dilute mineral acids Beilstein Reference: 1875653 Merck Reference: 14,3324 UN No.: 3077 MDL No.: MFCD00001758

Formula: C₁₃H₁₃N₃
Boiling Pt: >170 °C (dec.)
Melting Pt: 146…150 °C
Density: 1.130
Flash Pt: 170 °C (338 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001758
CAS Number: 102-06-7
EINECS: 203-002-1
UN: 3077
ADR: 9,III
Merck Index: 14,3324

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR