Benzofuran 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,3-Benzofuran, Benzo[b]furan, Benzo(b)furan, 1-Benzofuran, Coumarone

A15225-09 A15225-18
CAAAA15225-09EA 115.97 CAD
CAAAA15225-09 CAAAA15225-18
Benzofuran 99+%
Benzofuran

Purity: 99+% CAS No.: 271-89-6 Molecular Formula: C8H6O Formula Weight: 118.14 Storage Temperature: 2°C to 8°C Flash Point: 56°(133°F) Refractive Index: 1.5660 Odor: Aromatic Boiling Point: 173-175° Beilstein Reference: 107704 Merck Reference: 14,1088 UN No.: 1993 MDL No.: MFCD00005847

Formula: C₈H₆O
MW: 118.14 g/mol
Boiling Pt: 173…175 °C
Melting Pt: < –18 °C
Density: 1.095
Flash Pt: 56 °C (133 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00005847
CAS Number: 271-89-6
EINECS: 205-982-6
UN: 1993
ADR: 3,III
Merck Index: 14,1088

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR