Methyl-3,5-dihydroxybenzoate 98%

Supplier: Thermo Scientific Chemicals
Warning

A15224-09 A15224-18 A15224-30
CAAAA15224-09EA 62.57 CAD
CAAAA15224-09 CAAAA15224-18 CAAAA15224-30
Methyl-3,5-dihydroxybenzoate 98%
Methyl 3,5-dihydroxybenzoate

Purity: 98% CAS No.: 2150-44-9 Molecular Formula: C8H8O4 Formula Weight: 168.15 Melting Point: 164-168° Beilstein Reference: 2091651

Formula: C₈H₈O₄
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 164…168 °C
Storage Temperature: Ambient
MDL Number: MFCD00002289
CAS Number: 2150-44-9
EINECS: 218-426-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR