3,5-Dinitrobenzonitrile 98%

Supplier: Thermo Scientific Chemicals
Danger

A15223-06 A15223-14
CAAAA15223-06EA 85.64 CAD
CAAAA15223-06 CAAAA15223-14
3,5-Dinitrobenzonitrile 98%
3,5-Dinitrobenzonitrile

Purity: 98% CAS No.: 4110-35-4 Molecular Formula: C7H3N3O4 Formula Weight: 193.12 Melting Point: 127-130° UN No.: 3439 MDL No.: MFCD00007228

Formula: C₇H₃N₃O₄
Melting Pt: 127…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00007228
CAS Number: 4110-35-4
EINECS: 223-889-9
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR