2,5-Dimethylhexane 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15221-06
A15221-14
CAAAA15221-06EA
173.8
CAD
CAAAA15221-06
CAAAA15221-14
2,5-Dimethylhexane 99%
2,5-Dimethylhexane
Purity: 99%
CAS No.: 592-13-2
Molecular Formula: (CH3)2CHCH2CH2CH(CH3)2
Formula Weight: 114.23
Flash Point: 26°(80°F)
Refractive Index: 1.3920
Boiling Point: 107-108°
Beilstein Reference: 1696877
UN No.: 3295
MDL No.: MFCD00008951
Formula:
C₈H₁₈ MW: 114.23 g/mol Boiling Pt: 107…108 °C Melting Pt: –91 °C Density: 0.694 Flash Pt: 26 °C (80 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008951 CAS Number: 592-13-2 EINECS: 209-745-8 UN: 1262 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...