2,4-Difluorobenzyl bromide 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-(Bromomethyl)-2,4-difluorobenzene, α-Bromo-2,4-difluorotoluene
A15213-06
A15213-14
CAAAA15213-06EA
98.75
CAD
CAAAA15213-06
CAAAA15213-14
2,4-Difluorobenzyl bromide 98+%
2,4-Difluorobenzyl bromide
Purity: 98+%
CAS No.: 23915-07-3
Molecular Formula: C7H5BrF2
Formula Weight: 207.02
Flash Point: 85°(185°F)
Refractive Index: 1.5250
Boiling Point: 42-43°/1mm
Beilstein Reference: 4177539
UN No.: 1760
MDL No.: MFCD00011649
Formula:
C₇H₅BrF₂ MW: 207.02 g/mol Boiling Pt: 42…43 °C/1 mm Density: 1.613 Flash Pt: 85 °C (185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00011649 CAS Number: 23915-07-3 EINECS: 245-938-3 UN: 2924 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...