9-Fluorenylmethanol 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15212-06
A15212-14
A15212-22
CAAAA15212-06EA
95.59
CAD
CAAAA15212-06
CAAAA15212-14
CAAAA15212-22
9-Fluorenylmethanol 99%
9-Fluorenylmethanol
Purity: 99%
CAS No.: 24324-17-2
Molecular Formula: C14H12O
Formula Weight: 196.25
Melting Point: 102-106°
Beilstein Reference: 2330017
Formula:
C₁₄H₁₂O Boiling Pt: 337 °C (1013 hPa) Melting Pt: 102…106 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001139 CAS Number: 24324-17-2 EINECS: 246-167-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...