(2-Methoxyphenyl)acetic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Methoxyphenylacetic acid

A15211-14 A15211-09 A15211-18
CAAAAA15211-14EA 106.33 CAD
CAAAAA15211-14 CAAAA15211-09 CAAAA15211-18
(2-Methoxyphenyl)acetic acid 99%
(2-Methoxyphenyl)acetic acid

Purity: 99% CAS No.: 93-25-4 Molecular Formula: C9H10O3 Formula Weight: 166.18 Melting Point: 122-125° Beilstein Reference: 2047573

Formula: (C₆H₄OCH₃)CH₂COOH
Boiling Pt: 332 °C (1013 hPa)
Melting Pt: 122…125 °C
Storage Temperature: Ambient
MDL Number: MFCD00004321
CAS Number: 93-25-4
EINECS: 202-231-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR