ω,3-Dibromoacetophenone 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15210-06
A15210-14
CAAAA15210-03EA
79.7
CAD
CAAAA15210-03
CAAAA15210-06
CAAAA15210-14
ω,3-Dibromoacetophenone 97%
ω,3-Dibromoacetophenone
Purity: 97%
CAS No.: 18523-22-3
Molecular Formula: C8H6Br2O
Formula Weight: 277.95
Storage Temperature: 2°C to 8°C
Odor: Pungent
Beilstein Reference: 775786
UN No.: 3261
MDL No.: MFCD00082698
Formula:
C₈H₆Br₂O Melting Pt: 49…52 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00082698 CAS Number: 18523-22-3 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...