5-Chloro-2-thenoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Chlorothiophene-2-carboxylic acid, 5-Chloro-2-thiophenecarboxylic acid
A15209-06
A15209-14
A15209-22
CAAAA15209-06EA
46.45
CAD
CAAAA15209-06
CAAAA15209-14
CAAAA15209-22
5-Chloro-2-thenoic acid 98%
5-Chloro-2-thenoic acid
Purity: 98%
CAS No.: 24065-33-6
Molecular Formula: C5H3ClO2S
Formula Weight: 162.60
Melting Point: 149-152°
Beilstein Reference: 118361
Formula:
C₅H₃ClO₂S MW: 162.6 g/mol Melting Pt: 149…152 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041426 CAS Number: 24065-33-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...