3,4,5-Trimethoxytoluene 98%
Supplier: Thermo Scientific Chemicals
A15201-18
A15201-06
A15201-14
CAAAAA15201-18EA
176.96
CAD
CAAAAA15201-18
CAAAA15201-06
CAAAA15201-14
CAAAA15201-22
3,4,5-Trimethoxytoluene 98%
3,4,5-Trimethoxytoluene
Purity: 98%
CAS No.: 6443-69-2
Molecular Formula: C10H14O3
Formula Weight: 182.22
Flash Point: 74°(165°F)
Refractive Index: 1.5230
Melting Point: 34-38°
Boiling Point: 115-116°/4mm
Beilstein Reference: 1953791
MDL No.: MFCD00008397
Formula:
C₁₀H₁₄O₃ Boiling Pt: 115…116 °C/4 mm Melting Pt: 34…38 °C Density: 1.082 Flash Pt: 74 °C(165 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008397 CAS Number: 6443-69-2 EINECS: 229-239-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...