3,4-Dichloroaniline 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15199-18
A15199-30
A15199-0E
A15199-36
CAAAAA15199-18EA
41.55
CAD
CAAAAA15199-18
CAAAAA15199-30
CAAAA15199-0E
CAAAA15199-36
3,4-Dichloroaniline 98%
3,4-Dichloroaniline
Purity: 98%
CAS No.: 95-76-1
Molecular Formula: C6H5Cl2N
Formula Weight: 162.02
Flash Point: 135°(275°F)
Melting Point: 70-73°
Boiling Point: 272°
Beilstein Reference: 636837
Merck Reference: 14,3054
UN No.: 3442
MDL No.: MFCD00007768
Formula:
C₆H₅Cl₂N MW: 162.02 g/mol Boiling Pt: 272 °C Melting Pt: 70…73 °C Flash Pt: 135 °C (275 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007768 CAS Number: 95-76-1 EINECS: 202-448-4 UN: 3442 ADR: 6.1 II Merck Index: 14,3054 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...