DL-Serine 99%

Supplier: Thermo Scientific Chemicals

Synonyms: (±)-Serine, H-DL-Ser-OH, DL-2-Amino-3-hydroxypropionic acid

A15184-22 A15184-36
CAAAA15184-22EA 101.12 CAD
CAAAA15184-22 CAAAA15184-36
DL-Serine 99%
DL-Serine

Purity: 99% CAS No.: 302-84-1 Molecular Formula: C3H7NO3 Formula Weight: 105.09 Melting Point: ca 240° dec. Beilstein Reference: 1721402 Merck Reference: 14,8460 MDL No.: MFCD00064223

Formula: C₃H₇NO₃
MW: 105.09 g/mol
Boiling Pt: 395
Melting Pt: ∼ 240 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064223
CAS Number: 302-84-1
EINECS: 206-130-6
Merck Index: 14,8460

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR