(1-Hexadecyl)triphenylphosphonium bromide 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15180-06
A15180-22
A15180-09
CAAAAA15180-06EA
61.62
CAD
CAAAAA15180-06
CAAAAA15180-22
CAAAA15180-09
(1-Hexadecyl)triphenylphosphonium bromide 98+%
(1-Hexadecyl)triphenylphosphonium bromide
Purity: 98+%
CAS No.: 14866-43-4
Molecular Formula: CH3(CH2)15P(C6H5)3Br
Formula Weight: 567.64
Melting Point: 99-101°
Beilstein Reference: 3582592
Formula:
H₃C(CH₂)₁₅P(C₆H₅)₃Br MW: 567.63 g/mol Melting Pt: 99…101 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051858 CAS Number: 14866-43-4 EINECS: 238-939-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...