3,5-Difluorophenol 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15177-03
A15177-06
A15177-14
CAAAA15177-03EA
48.82
CAD
CAAAA15177-03
CAAAA15177-06
CAAAA15177-14
3,5-Difluorophenol 98+%
3,5-Difluorophenol
Purity: 98+%
CAS No.: 2713-34-0
Molecular Formula: C6H4F2O
Formula Weight: 130.09
Flash Point: 70°(158°F)
Odor: Phenol-like
Boiling Point: 65-68°/1mm
Beilstein Reference: 2078616
UN No.: 3261
MDL No.: MFCD00002255
Formula:
C₆H₄F₂O MW: 130.09 g/mol Boiling Pt: 65…68 °C/1 mm Melting Pt: 51…55 °C Density: 1.566 g/cm³ (20 °C) Flash Pt: 70 °C (158 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002255 CAS Number: 2713-34-0 UN: 2925 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...