trans-2-Methoxycinnamic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15173-06
A15173-14
A15173-22
CAAAA15173-06EA
46.14
CAD
CAAAA15173-06
CAAAA15173-14
CAAAA15173-22
trans-2-Methoxycinnamic acid 98+%
trans-2-Methoxycinnamic acid
Purity: 98+%
CAS No.: 1011-54-7
Molecular Formula: C10H10O3
Formula Weight: 178.19
Melting Point: 183-186°
Beilstein Reference: 2209712
Formula:
C₁₀H₁₀O₃ Melting Pt: 183…186 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064238 CAS Number: 1011-54-7 EINECS: 213-785-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...