3,5-Dichlorobenzoic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

A15168-06 A15168-14 A15168-22
CAAAA15168-06EA 67.47 CAD
CAAAA15168-06 CAAAA15168-14 CAAAA15168-22
3,5-Dichlorobenzoic acid 99%
3,5-Dichlorobenzoic acid

Purity: 99% CAS No.: 51-36-5 Molecular Formula: C7H4Cl2O2 Formula Weight: 191.01 Melting Point: 185-187° Beilstein Reference: 2044776

Formula: C₇H₄Cl₂O₂
MW: 191.01 g/mol
Boiling Pt: 309 °C (1013 hPa)
Melting Pt: 185…187 °C
Storage Temperature: Ambient
MDL Number: MFCD00002494
CAS Number: 51-36-5
EINECS: 200-092-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR