1-Bromohexane 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Hexyl bromide
A15166-22
A15166-0C
A15166-0E
A15166-36
CAAAAA15166-22EA
40.29
CAD
CAAAAA15166-22
CAAAA15166-0C
CAAAA15166-0E
CAAAA15166-36
1-Bromohexane 99%
1-Bromohexane
Purity: 99%
CAS No.: 111-25-1
Molecular Formula: CH3(CH2)5Br
Formula Weight: 165.08
Flash Point: 50°(122°F)
Refractive Index: 1.4475
Odor: Pleasant
Melting Point: -85°
Boiling Point: 156°
Beilstein Reference: 1731290
UN No.: 1993
MDL No.: MFCD00000271
Formula:
BrH₂C(CH₂)₄CH₃ MW: 165.07 g/mol Boiling Pt: 156 °C Melting Pt: –85 °C Density: 1.176 Flash Pt: 50 °C (122 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000271 CAS Number: 111-25-1 EINECS: 203-850-2 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...