Phenylpropiolic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15163-14
A15163-09
A15163-18
CAAAAA15163-14EA
279.66
CAD
CAAAAA15163-14
CAAAA15163-09
CAAAA15163-18
Phenylpropiolic acid 98+%
Phenylpropiolic acid
Purity: 98+%
CAS No.: 637-44-5
Molecular Formula: C6H5C≡CCO2H
Formula Weight: 146.15
Melting Point: 135-138°
Beilstein Reference: 742587
Formula:
C₉H₆O₂ Boiling Pt: 294 °C (1013 hPa) Melting Pt: 135…138 °C Density: 1.28 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004361 CAS Number: 637-44-5 EINECS: 211-285-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...