4-Isopropylcinnamic acid predominantly trans 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(4-Isopropylphenyl)acrylic acid

A15156-06 A15156-14
CAAAA15156-06EA 122.13 CAD
CAAAA15156-06 CAAAA15156-14
4-Isopropylcinnamic acid predominantly trans 98+%
4-Isopropylcinnamic acid

Purity: 98+% CAS No.: 3368-21-6 Molecular Formula: C12H14O2 Formula Weight: 190.24 Melting Point: 157-161° Beilstein Reference: 2046679

Formula: C₁₂H₁₄O₂
MW: 190.24
Melting Pt: 157…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00016544
CAS Number: 3368-21-6
EINECS: 222-138-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR