Methyl-3-nitrobenzoate 98+%
Supplier: Thermo Scientific Chemicals
A15155-18
A15155-30
CAAAA15155-18EA
60.99
CAD
CAAAA15155-18
CAAAA15155-30
Methyl-3-nitrobenzoate 98+%
Methyl 3-nitrobenzoate
Purity: 98+%
CAS No.: 618-95-1
Molecular Formula: C8H7NO4
Formula Weight: 181.15
Melting Point: 77-80°
Boiling Point: 279°
MDL No.: MFCD00007250
Formula:
C₈H₇NO₄ Boiling Pt: 279 °C Melting Pt: 77…80 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007250 CAS Number: 618-95-1 EINECS: 210-573-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...