4-Chloro-α,α,α-trifluorotoluene 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Chloro-4-(trifluoromethyl)benzene, 4-Chlorobenzotrifluoride
A15154-0E
A15154-22
A15154-36
CAAAA15154-0EEA
535.62
CAD
CAAAA15154-0E
CAAAA15154-22
CAAAA15154-36
4-Chloro-α,α,α-trifluorotoluene 98+%
4-Chloro-α,α,α-trifluorotoluene
Purity: 98+%
CAS No.: 98-56-6
Molecular Formula: C7H4ClF3
Formula Weight: 180.56
Flash Point: 47°(116°F)
Refractive Index: 1.4460
Melting Point: -36°
Boiling Point: 136-138°
Beilstein Reference: 510203
Merck Reference: 14,2126
UN No.: 2234
MDL No.: MFCD00000627
Formula:
C₇H₄ClF₃ MW: 180.56 g/mol Boiling Pt: 136…138 °C Melting Pt: –36 °C Density: 1.346 Flash Pt: 47 °C (116 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000627 CAS Number: 98-56-6 EINECS: 202-681-1 UN: 2234 ADR: 3,III Merck Index: 14,2126 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...