2-Isopropylnaphthalene 96%, Technical Grade

Supplier: Thermo Scientific Chemicals

A15153-06 A15153-14
CAAAA15153-06EA 131.93 CAD
CAAAA15153-06 CAAAA15153-14
2-Isopropylnaphthalene 96%, Technical Grade
2-Isopropylnaphthalene

Purity: 96% CAS No.: 2027-17-0 Molecular Formula: C13H14 Formula Weight: 170.26 Flash Point: 122°(252°F) Refractive Index: 1.5870 Melting Point: 14° Boiling Point: 268° MDL No.: MFCD00014322

Formula: C₁₃H₁₄
Boiling Pt: 268 °C
Melting Pt: 14 °C
Density: 0.975
Flash Pt: 122 °C(252 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014322
CAS Number: 2027-17-0
EINECS: 217-976-0
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR