1-Bromohexadecane 98%
Supplier: Thermo Scientific Chemicals
A15147-14
A15147-0B
A15147-30
CAAAAA15147-14EA
38.24
CAD
CAAAAA15147-14
CAAAA15147-0B
CAAAA15147-30
1-Bromohexadecane 98%
1-Bromohexadecane
Purity: 98%
CAS No.: 112-82-3
Molecular Formula: CH3(CH2)15Br
Formula Weight: 305.35
Flash Point: 176°(348°F)
Refractive Index: 1.4620
Odor: Odorless
Melting Point: 16-18°
Boiling Point: 336°
Beilstein Reference: 773989
MDL No.: MFCD00000230
Formula:
BrH₂C(CH₂)₁₄CH₃ Boiling Pt: 336 °C Melting Pt: 16…18 °C Density: 1.000 Flash Pt: 176 °C(348 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000230 CAS Number: 112-82-3 EINECS: 204-008-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...