Propiophenone 99%
Supplier: Thermo Scientific Chemicals
Warning
|
A15140-0E
A15140-22
A15140-36
CAAAA15140-0EEA
352.34
CAD
CAAAA15140-0E
CAAAA15140-22
CAAAA15140-36
Propiophenone 99%
Propiophenone
Purity: 99%
CAS No.: 93-55-0
Molecular Formula: C9H10O
Formula Weight: 134.18
Flash Point: 87°(188°F)
Refractive Index: 1.5260
Melting Point: 17-18°
Boiling Point: 217-218°
Beilstein Reference: 606215
Merck Reference: 14,7830
MDL No.: MFCD00009309
Formula:
C₆H₅C(O)CH₂CH₃ Boiling Pt: 217…218 °C Melting Pt: 17…18 °C Density: 1.008 Flash Pt: 87 °C(188 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009309 CAS Number: 93-55-0 EINECS: 202-257-6 Merck Index: 14,7830 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...