HMDS (1,1,1,3,3,3-hexamethyldisilazane) 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Hexamethyldisilazane

A15139-0F A15139-AE A15139-AP
CAAAAA15139-ACEA 33.65 CAD
CAAAAA15139-AC CAAAA15139-0F CAAAA15139-AE CAAAA15139-AP
HMDS (1,1,1,3,3,3-hexamethyldisilazane) 98+%
HMDS (1,1,1,3,3,3-Hexamethyldisilazane)

Purity: 98+% CAS No.: 999-97-3 Molecular Formula: C6H19NSi2 Formula Weight: 161.40 Flash Point: 8°(46°F) Physical Form: Clear liquid Density: 0.774 Refractive Index: 1.4080 Sensitivity: Moisture Sensitive Appearance: Colorless Odor: Odorless Melting Point: -78° Boiling Point: 126° Beilstein Reference: 635752 Merck Reference: 14,4689 UN No.: 3286 MDL No.: MFCD00008259

Formula: (CH₃)₃SiNHSi(CH₃)₃
MW: 161.39 g/mol
Boiling Pt: 126 °C
Melting Pt: –78 °C
Density: 0.774
Flash Pt: 8 °C (46 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008259
CAS Number: 999-97-3
EINECS: 213-668-5
UN: 3286
ADR: 3,II
REACH: 01-2119438176-38
Merck Index: 14,4689
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce35a38a4179292c4ff37b2f3093018738545e6e&vid=1693ad54a45ca3ae50a06056893e8438d1002549&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR