4-Chlorobenzoic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
p-Chlorobenzoic acid, Chlorodracyclic acid, Chlorodracylic acid
A15135-0B
A15135-30
CAAAA15135-0BEA
311.26
CAD
CAAAA15135-0B
CAAAA15135-30
4-Chlorobenzoic acid 98+%
4-Chlorobenzoic acid
Purity: 98+%
CAS No.: 74-11-3
Molecular Formula: C7H5ClO2
Formula Weight: 156.57
Flash Point: 238°(460°F)
Odor: Odorless
Boiling Point: 275°
Beilstein Reference: 907196
Merck Reference: 14,2125
MDL No.: MFCD00002531
Formula:
C₇H₅ClO₂ MW: 156.57 g/mol Boiling Pt: 275 °C Melting Pt: 239…243 °C Density: 1.54 Flash Pt: 238 °C(460 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002531 CAS Number: 74-11-3 EINECS: 200-805-9 Merck Index: 14,2125 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...