p-Tolylacetic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15129-06
A15129-14
A15129-22
CAAAA15129-06EA
44.24
CAD
CAAAA15129-06
CAAAA15129-14
CAAAA15129-22
p-Tolylacetic acid 99%
p-Tolylacetic acid
Purity: 99%
CAS No.: 622-47-9
Molecular Formula: C9H10O2
Formula Weight: 150.18
Melting Point: 90-93°
Boiling Point: 265-267°
MDL No.: MFCD00004353
Formula:
(C₆H₄(CH₃))CH₂COOH Boiling Pt: 265…267 °C Melting Pt: 90…93 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004353 CAS Number: 622-47-9 EINECS: 210-738-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...