2'-Fluoroacetophenone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15128-06
A15128-14
A15128-22
CAAAA15128-06EA
63.52
CAD
CAAAA15128-06
CAAAA15128-14
CAAAA15128-22
2'-Fluoroacetophenone 97%
2'-Fluoroacetophenone
Purity: 97%
CAS No.: 445-27-2
Molecular Formula: C8H7FO
Formula Weight: 138.14
Flash Point: 61°(141°F)
Refractive Index: 1.5075
Boiling Point: 82-83°/10mm
Beilstein Reference: 2041344
MDL No.: MFCD00000320
Formula:
C₈H₇FO Boiling Pt: 82…83 °C/10 mm Density: 1.137 Flash Pt: 61 °C(141 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000320 CAS Number: 445-27-2 EINECS: 207-156-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...