4-Bromoaniline 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15126-0B
A15126-18
A15126-30
CAAAA15126-0BEA
624.1
CAD
CAAAA15126-0B
CAAAA15126-18
CAAAA15126-30
4-Bromoaniline 98+%
4-Bromoaniline
Purity: 98+%
CAS No.: 106-40-1
Molecular Formula: C6H6BrN
Formula Weight: 172.03
Physical Form: Cryst solid
Odor: Characteristic
Boiling Point: 222-224°
Beilstein Reference: 742031
Merck Reference: 14,1404
UN No.: 2811
MDL No.: MFCD00007822
Formula:
C₆H₆BrN Boiling Pt: 222…224 °C Melting Pt: 60…65 °C Density: 1.799 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007822 CAS Number: 106-40-1 EINECS: 203-393-9 UN: 2811 ADR: 6.1,III Merck Index: 14,1404 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...