Adipohydrazide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Adipyl hydrazide, Adipic acid dihydrazide, Adipic dihydrazide
A15119-36
A15119-14
A15119-22
CAAAAA15119-36EA
820.02
CAD
CAAAAA15119-36
CAAAA15119-14
CAAAA15119-22
Adipohydrazide 97%
Adipohydrazide
Purity: 97%
CAS No.: 1071-93-8
Molecular Formula: H2NNHCO(CH2)4CONHNH2
Formula Weight: 174.20
Odor: Odorless
Melting Point: 180-182°
Beilstein Reference: 973863
MDL No.: MFCD00007614
Formula:
C₆H₁₄N₄O₂ Boiling Pt: 446 °C (1013 hPa) Melting Pt: 180…182 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007614 CAS Number: 1071-93-8 EINECS: 213-999-5 UN: 3077 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...