3,5-Dimethoxybenzaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

A15109-06 A15109-14 A15109-22
CAAAA15109-06EA 124.03 CAD
CAAAA15109-06 CAAAA15109-14 CAAAA15109-22
3,5-Dimethoxybenzaldehyde 98%
3,5-Dimethoxybenzaldehyde

Purity: 98% CAS No.: 7311-34-4 Molecular Formula: C9H10O3 Formula Weight: 166.18 Flash Point: 110°(230°F) Melting Point: 44-48° Boiling Point: 115-118°/1mm Beilstein Reference: 2042374 MDL No.: MFCD00003366

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 115…118 °C/1 mm
Melting Pt: 44…48 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003366
CAS Number: 7311-34-4
EINECS: 230-772-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR