2-Chloro-5-nitroaniline 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15102-22
A15102-36
CAAAA15102-22EA
55.3
CAD
CAAAA15102-22
CAAAA15102-36
2-Chloro-5-nitroaniline 98%
2-Chloro-5-nitroaniline
Purity: 98%
CAS No.: 6283-25-6
Molecular Formula: C6H5ClN2O2
Formula Weight: 172.57
Flash Point: 191°(375°F)
Odor: Weak, characteristic
Beilstein Reference: 2208878
UN No.: 2237
MDL No.: MFCD00007668
Formula:
C₆H₅ClN₂O₂ Melting Pt: 117…121 °C Flash Pt: 191 °C (375 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007668 CAS Number: 6283-25-6 EINECS: 228-498-7 UN: 2237 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...