4,4'-Dihydroxybenzophenone 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Bis(4-hydroxyphenyl) ketone
A15097-06
A15097-14
A15097-22
CAAAA15097-06EA
99.7
CAD
CAAAA15097-06
CAAAA15097-14
CAAAA15097-22
4,4'-Dihydroxybenzophenone 98+%
4,4'-Dihydroxybenzophenone
Purity: 98+%
CAS No.: 611-99-4
Molecular Formula: C13H10O3
Formula Weight: 214.22
Melting Point: 221-225°
Beilstein Reference: 1874572
Formula:
C₁₃H₁₀O₃ MW: 214.22 g/mol Boiling Pt: 375 Melting Pt: 221…225 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002358 CAS Number: 611-99-4 EINECS: 210-288-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...