4-Hydroxybenzophenone 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (4-Hydroxyphenyl)(phenyl)methanone, p-Hydroxybenzophenone

A15096-0B A15096-18 A15096-30
CAAAA15096-0BEA 1260.84 CAD
CAAAA15096-0B CAAAA15096-18 CAAAA15096-30
4-Hydroxybenzophenone 98%
4-Hydroxybenzophenone

Purity: 98% CAS No.: 1137-42-4 Molecular Formula: C13H10O2 Formula Weight: 198.22 Melting Point: 132-135° Boiling Point: 260-262°/24mm MDL No.: MFCD00002355

Formula: C₁₃H₁₀O₂
MW: 198.22 g/mol
Boiling Pt: 260…262 °C (24 mmHg)
Melting Pt: 132…135 °C
Storage Temperature: Ambient
MDL Number: MFCD00002355
CAS Number: 1137-42-4
EINECS: 214-507-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR