3,5-Dibromo-1-fluorobenzene 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15078-22
A15078-14
CAAAAA15078-22EA
579.86
CAD
CAAAAA15078-22
CAAAA15078-06
CAAAA15078-14
3,5-Dibromo-1-fluorobenzene 98+%
3,5-Dibromo-1-fluorobenzene
Purity: 98+%
CAS No.: 1435-51-4
Molecular Formula: C6H3Br2F
Formula Weight: 253.91
Flash Point:
Formula:
C₆H₃Br₂F Boiling Pt: 204…206 °C Density: 2,018 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00061119 CAS Number: 1435-51-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...