2,5-Dibromobenzoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15077-06
A15077-14
CAAAA15077-06EA
108.39
CAD
CAAAA15077-06
CAAAA15077-14
2,5-Dibromobenzoic acid 98%
2,5-Dibromobenzoic acid
Purity: 98%
CAS No.: 610-71-9
Molecular Formula: C7H4Br2O2
Formula Weight: 279.93
Melting Point: 153-157°
Beilstein Reference: 1868193
Formula:
C₇H₄Br₂O₂ MW: 279.92 Melting Pt: 153…157 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016494 CAS Number: 610-71-9 EINECS: 210-234-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...