α,3-Dibromotoluene 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Bromobenzyl bromide, 1-Bromo-3-(bromomethyl)benzene

A15075-09 A15075-18 A15075-30
CAAAA15075-09EA 93.38 CAD
CAAAA15075-09 CAAAA15075-18 CAAAA15075-30
α,3-Dibromotoluene 99%
α,3-Dibromotoluene

Purity: 99% CAS No.: 823-78-9 Molecular Formula: C7H6Br2 Formula Weight: 249.94 Flash Point: 110°(230°F) Melting Point: 40-44° Boiling Point: 128-130°/12mm Beilstein Reference: 2078683 UN No.: 1759 MDL No.: MFCD00000176

Formula: C₇H₆Br₂
Boiling Pt: 128…130 °C/12 mm
Melting Pt: 40…44 °C
Density: 1.560
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000176
CAS Number: 823-78-9
EINECS: 212-519-1
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR