Phthalonitrile 98%

Supplier: Thermo Scientific Chemicals
Danger

A15071-22 A15071-0B A15071-30
CAAAAA15071-22EA 50.72 CAD
CAAAAA15071-22 CAAAA15071-0B CAAAA15071-30
Phthalonitrile 98%
Phthalonitrile

Purity: 98% CAS No.: 91-15-6 Molecular Formula: C8H4N2 Formula Weight: 128.13 Flash Point: 162°(323°F) Melting Point: 139-141° Beilstein Reference: 775028 UN No.: 3439 MDL No.: MFCD00001771

Formula: C₈H₄N₂
Boiling Pt: 305 °C (1013 hPa)
Melting Pt: 139…141 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 162 °C (323 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001771
CAS Number: 91-15-6
EINECS: 202-044-8
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR