(3-Methoxyphenyl)acetic acid ethyl ester 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Ethyl (3-methoxyphenyl)acetate, Ethyl 2-(3-methoxyphenyl)acetate

A15070-06 A15070-14
CAAAA15070-06EA 219.62 CAD
CAAAA15070-06 CAAAA15070-14
(3-Methoxyphenyl)acetic acid ethyl ester 98%
(3-Methoxyphenyl)acetic acid ethyl ester

Purity: 98% CAS No.: 35553-92-5 Molecular Formula: C11H4O3 Formula Weight: 194.23 Boiling Point: 109-110°/1mm MDL No.: MFCD00026897

Formula: C₁₁H₁₄O₃
MW: 194.23 g/mol
Boiling Pt: 109…110 °C/1 mm
Storage Temperature: Ambient
MDL Number: MFCD00026897
CAS Number: 35553-92-5
EINECS: 252-614-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR