2-Bromobutane 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15069-0B
A15069-30
CAAAA15069-0BEA
252.8
CAD
CAAAA15069-0B
CAAAA15069-30
2-Bromobutane 98%
2-Bromobutane
Purity: 98%
CAS No.: 78-76-2
Molecular Formula: C4H9Br
Formula Weight: 137.03
Flash Point: 13°(55°F)
Physical Form: Liquid
Density: 1.260
Refractive Index: 1.4369
Appearance: Colorless
Odor: Pleasant
Melting Point: -112°
Boiling Point: 91-92°
Beilstein Reference: 505949
Merck Reference: 14,1554
UN No.: 2339
MDL No.: MFCD00000156
Formula:
H₃CBrCHCH₂CH₃ Boiling Pt: 91…92 °C Melting Pt: –112 °C Density: 1.260 Flash Pt: 13 °C (55 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000156 CAS Number: 78-76-2 EINECS: 201-140-7 UN: 2339 ADR: 3,II Merck Index: 14,1554 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...