Ethyl propiolate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15068-06
A15068-14
A15068-22
CAAAA15068-06EA
76.63
CAD
CAAAA15068-06
CAAAA15068-14
CAAAA15068-22
Ethyl propiolate 99%
Ethyl propiolate
Purity: 99%
CAS No.: 623-47-2
Molecular Formula: C5H6O2
Formula Weight: 98.10
Storage Temperature: 2°C to 8°C
Flash Point: 23°(73°F)
Refractive Index: 1.4120
Odor: Pungent
Melting Point: 9°
Boiling Point: 118-120°
Beilstein Reference: 878250
UN No.: 3272
MDL No.: MFCD00009184
Formula:
C₅H₆O₂ Boiling Pt: 118…120 °C Melting Pt: 9 °C Density: 0.963 Flash Pt: 23 °C (73 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00009184 CAS Number: 623-47-2 EINECS: 210-795-8 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...