2-Iodopropane 98+% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
Stabilization:
Stabilised with copper
A15066-14
A15066-22
A15066-36
CAAAAA15066-14EA
52.46
CAD
CAAAAA15066-14
CAAAA15066-22
CAAAA15066-36
2-Iodopropane 98+% stabilized
2-Iodopropane
Purity: 98+%
CAS No.: 75-30-9
Molecular Formula: (CH3)2CHI
Formula Weight: 169.99
Flash Point: 42°(107°F)
Physical Form: Liquid
Density: 1.701
Refractive Index: 1.4982
Sensitivity: Light Sensitive
Odor: Recognizable
Melting Point: -90°
Boiling Point: 88-90°
Beilstein Reference: 1098244
Merck Reference: 14,5214
UN No.: 2392
MDL No.: MFCD00001071
Formula:
C₃H₇I Boiling Pt: 88…90 °C Melting Pt: –90 °C Density: 1.701 Flash Pt: 42 °C (107 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001071 CAS Number: 75-30-9 EINECS: 200-859-3 UN: 2392 ADR: 3,III Merck Index: 14,5214 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...