2-Bromobenzaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15065-14
A15065-22
A15065-36
CAAAA15065-14EA
111.86
CAD
CAAAA15065-14
CAAAA15065-22
CAAAA15065-36
2-Bromobenzaldehyde 98%
2-Bromobenzaldehyde
Purity: 98%
CAS No.: 6630-33-7
Molecular Formula: C7H5BrO
Formula Weight: 185.03
Flash Point: 95°(203°F)
Refractive Index: 1.5960
Odor: Characteristic
Melting Point: 18-21°
Boiling Point: 228-230°
Beilstein Reference: 636132
MDL No.: MFCD00003300
Formula:
C₇H₅BrO Boiling Pt: 228…230 °C Melting Pt: 18…21 °C Density: 1,597 Flash Pt: 95 °C(203 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003300 CAS Number: 6630-33-7 EINECS: 229-622-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...