(4-Methoxyphenyl)acetonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4-Methoxybenzyl cyanide, 4-Methoxyphenylacetonitrile
A15056-14
A15056-22
CAAAA15056-14EA
50.24
CAD
CAAAA15056-14
CAAAA15056-22
(4-Methoxyphenyl)acetonitrile 98%
(4-Methoxyphenyl)acetonitrile
Purity: 98%
CAS No.: 104-47-2
Molecular Formula: C9H9NO
Formula Weight: 147.18
Flash Point: 117°(242°F)
Refractive Index: 1.5300
Melting Point: 8°
Boiling Point: 285-287°
Beilstein Reference: 509162
UN No.: 3276
MDL No.: MFCD00001919
Formula:
C₉H₉NO Boiling Pt: 285…287 °C Melting Pt: 8 °C Density: 1.085 Flash Pt: 117 °C (242 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001919 CAS Number: 104-47-2 EINECS: 203-206-0 UN: 3276 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...